This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02637132_m1
See other PCMTD2 GE Assays ›
Gene Symbol:
PCMTD2
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 2
Gene Aliases:
-
Chromosome Location:
Chr.24: 47649520 - 47672218 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02637132_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCMTD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005635329.3 XP_005635386.1 6 - 7 926 125
XM_022409013.1 XP_022264721.1 6 - 7 926 125
XM_022409014.1 XP_022264722.1 6 - 7 1067 125
XM_022409015.1 XP_022264723.1 6 - 7 1006 125
XM_022409016.1 XP_022264724.1 5 - 6 941 125

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0