This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02637090_m1
See other ZNF567 GE Assays ›
Gene Symbol:
ZNF567
Gene Name:
zinc finger protein 567
Gene Aliases:
ZNF780A, ZNF780B
Chromosome Location:
Chr.1: 116337603 - 116382287 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02637090_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF567
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014115501.2 XP_013970976.2 2 - 3 180 80
XM_022403715.1 XP_022259423.1 3 - 4 282 80
XM_022403718.1 XP_022259426.1 3 - 4 312 80
XM_022403719.1 XP_022259427.1 3 - 4 278 80
XM_022403722.1 XP_022259430.1 2 - 3 117 80
XM_022403730.1 XP_022259438.1 3 - 4 282 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_014115501.2, XM_022403715.1, XM_022403718.1, XM_022403719.1, XM_022403722.1, XM_022403730.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0