This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02636927_m1
See other DNAJB4 GE Assays ›
Gene Symbol:
DNAJB4
Gene Name:
DnaJ heat shock protein family (Hsp40) member B4
Gene Aliases:
-
Chromosome Location:
Chr.6: 68878823 - 68923306 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02636927_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNAJB4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005622042.3 XP_005622099.1 3 - 4 930 155
XM_022420604.1 XP_022276312.1 2 - 3 1140 155
XM_022420605.1 XP_022276313.1 3 - 4 934 155
XM_022420606.1 XP_022276314.1 2 - 3 657 155
XM_022420607.1 XP_022276315.1 3 - 4 811 155
XM_022420609.1 XP_022276317.1 2 - 3 668 155
XM_022420610.1 XP_022276318.1 2 - 3 666 155
XM_022420611.1 XP_022276319.1 2 - 3 730 155
XM_022420612.1 XP_022276320.1 2 - 3 669 155
XM_862584.5 XP_867677.2 3 - 4 1164 155

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0