This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02635652_m1
See other LOC478407 GE Assays ›
Gene Symbol:
LOC478407 LOC100856622
Gene Name:
protein downstream neighbor of Son
protein downstream neighbor of Son
Gene Aliases:
DONSON
-
Chromosome Location:
Chr.31: 29097512 - 29109088 on Build CanFam3.1
Chromosome Location:
Chr.31: 28192338 - 28209512 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf02635652_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC478407
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014109694.2 XP_013965169.1 8 - 9 1421 103
Gene Symbol: LOC100856622
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005638851.3 XP_005638908.1 6 - 7 1121 103
XM_005638852.2 XP_005638909.1 6 - 7 1121 103
XM_022413295.1 XP_022269003.1 6 - 7 1121 103
XM_022413296.1 XP_022269004.1 6 - 7 1121 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0