This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02633865_m1
See other SEC14L2 GE Assays ›
Gene Symbol:
SEC14L2
Gene Name:
SEC14 like lipid binding 2
Gene Aliases:
-
Chromosome Location:
Chr.26: 23542555 - 23563579 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02633865_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEC14L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005636443.3 XP_005636500.2 2 - 3 195 62
XM_022410423.1 XP_022266131.1 2 - 3 198 62
XM_022410424.1 XP_022266132.1 2 - 3 196 62
XM_022410425.1 XP_022266133.1 3 - 4 364 62
XM_022410426.1 XP_022266134.1 3 - 4 279 62
XM_022410427.1 XP_022266135.1 3 - 4 259 62
XM_534734.6 XP_534734.3 3 - 4 279 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005636443.3, XM_022410423.1, XM_022410424.1, XM_022410427.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0