This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02633639_mH
See other LOC477516 GE Assays ›
Gene Symbol:
LOC477516
Gene Name:
calcium binding protein 1
Gene Aliases:
CABP1
Chromosome Location:
Chr.26: 16492484 - 16523265 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02633639_mH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC477516
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022410295.1 XP_022266003.1 8 - 9 1107 104
XM_022410296.1 XP_022266004.1 7 - 8 871 104
XM_022410297.1 XP_022266005.1 3 - 4 798 104
XM_022410298.1 XP_022266006.1 3 - 4 230 104
XM_534711.6 XP_534711.3 4 - 5 2972 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header