This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02633638_m1
See other LOC477516 GE Assays ›
Gene Symbol:
LOC477516
Gene Name:
calcium binding protein 1
Gene Aliases:
CABP1
Chromosome Location:
Chr.26: 16492484 - 16523265 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02633638_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC477516
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022410295.1 XP_022266003.1 7 - 8 966 66
XM_022410296.1 XP_022266004.1 6 - 7 730 66
XM_022410297.1 XP_022266005.1 2 - 3 657 66
XM_022410298.1 XP_022266006.1 2 - 3 89 66
XM_534711.6 XP_534711.3 3 - 4 2831 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0