This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02633136_g1
See other DDA1 GE Assays ›
Gene Symbol:
DDA1
Gene Name:
DET1 and DDB1 associated 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 45440046 - 45448564 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02633136_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DDA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014122095.2 XP_013977570.1 2 - 3 199 62
XM_022406896.1 XP_022262604.1 2 - 3 251 62
XM_022406897.1 XP_022262605.1 2 - 3 136 62
XM_022406898.1 XP_022262606.1 2 - 3 124 62
XM_022406899.1 XP_022262607.1 2 - 3 449 62
XM_533880.6 XP_533880.1 2 - 3 211 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0