This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02630490_m1
See other FGFR1OP2 GE Assays ›
Gene Symbol:
FGFR1OP2
Gene Name:
FGFR1 oncogene partner 2
Gene Aliases:
-
Chromosome Location:
Chr.27: 20672166 - 20704530 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02630490_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGFR1OP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197313.1 NP_001184242.1 1 - 2 156 83
XM_005636710.3 XP_005636767.1 2 - 3 411 83
XM_005636711.3 XP_005636768.1 2 - 3 411 83
XM_005636712.3 XP_005636769.1 2 - 3 411 83
GenBank mRNA GU811150.1 - 1 - 2 156 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0