This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02629695_m1
See other S100PBP GE Assays ›
Gene Symbol:
S100PBP
Gene Name:
S100P binding protein
Gene Aliases:
-
Chromosome Location:
Chr.2: 68495783 - 68535847 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02629695_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100PBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005617625.3 XP_005617682.1 4 - 5 1171 135
XM_005617629.3 XP_005617686.1 3 - 4 1057 135
XM_005617631.3 XP_005617688.1 4 - 5 1171 135
XM_022411866.1 XP_022267574.1 4 - 5 1070 135
XM_022411867.1 XP_022267575.1 4 - 5 1070 135
XM_022411869.1 XP_022267577.1 4 - 5 1072 135
XM_535323.6 XP_535323.3 4 - 5 1168 135

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0