This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02628290_m1
See other PTHLH GE Assays ›
Gene Symbol:
PTHLH
Gene Name:
parathyroid hormone like hormone
Gene Aliases:
PTHRP
Chromosome Location:
Chr.27: 19781557 - 19795078 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02628290_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTHLH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003303.1 NP_001003303.1 3 - 4 690 81
XM_022410845.1 XP_022266553.1 3 - 4 1263 81
XM_022410846.1 XP_022266554.1 4 - 5 1148 81
XM_022410847.1 XP_022266555.1 3 - 4 1097 81
XM_022410848.1 XP_022266556.1 4 - 5 982 81
XM_022410849.1 XP_022266557.1 3 - 4 881 81
GenBank mRNA U15593.1 - 3 - 4 690 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

peptide hormone

Gene Ontology Categories:

Function(s) Process(es)

bone mineralization
peptide hormone receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0