This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02628044_m1
See other PLA2G1B GE Assays ›
Gene Symbol:
PLA2G1B
Gene Name:
phospholipase A2 group IB
Gene Aliases:
-
Chromosome Location:
Chr.26: 16248417 - 16257163 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02628044_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLA2G1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003320.1 NP_001003320.1 2 - 3 224 61
GenBank mRNA M35301.1 - 2 - 3 224 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

innate immune response in mucosa
lipid catabolic process
antibacterial humoral response
phosphatidylcholine metabolic process
phosphatidylglycerol metabolic process
arachidonic acid secretion
defense response to Gram-positive bacterium
antimicrobial humoral immune response mediated by antimicrobial peptide
positive regulation of glomerular visceral epithelial cell apoptotic process
calcium ion binding
bile acid binding
calcium-dependent phospholipase A2 activity
phospholipase A2 activity (consuming 1,2-dipalmitoylphosphatidylcholine)
phospholipase A2 activity consuming 1,2-dioleoylphosphatidylethanolamine)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0