This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02627691_m1
See other EMR4 GE Assays ›
Gene Symbol:
EMR4
Gene Name:
EGF-like module containing, mucin-like, hormone receptor-like sequence 4
Gene Aliases:
EMR4P
Chromosome Location:
Chr.20: 53342233 - 53385263 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02627691_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EMR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038665.2 NP_001033754.2 9 - 10 1005 69
XM_005632346.3 XP_005632403.1 10 - 11 1136 69
GenBank mRNA DQ227278.1 - 9 - 10 1005 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 G-protein coupled receptor Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0