This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02626290_m1
See other CD40 GE Assays ›
Gene Symbol:
CD40
Gene Name:
CD40 molecule
Gene Aliases:
TNFRSF5
Chromosome Location:
Chr.24: 33352992 - 33383488 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02626290_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD40
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002982.1 NP_001002982.1 6 - 7 757 115
XM_005634663.3 XP_005634720.1 6 - 7 654 115
XM_005634664.3 XP_005634721.1 6 - 7 654 115
XM_005634665.3 XP_005634722.1 6 - 7 654 115
XM_005634666.3 XP_005634723.1 7 - 8 893 115
XM_005634667.3 XP_005634724.1 7 - 8 871 115
XM_022408802.1 XP_022264510.1 7 - 8 888 115
XM_022408803.1 XP_022264511.1 6 - 7 767 115
XM_022408804.1 XP_022264512.1 6 - 7 533 115
XM_022408805.1 XP_022264513.1 5 - 6 475 115
GenBank mRNA AY333789.1 - 6 - 7 757 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

B cell activation
regulation of immune response
signaling receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0