This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02624257_m1
See other SLAMF1 GE Assays ›
Gene Symbol:
SLAMF1
Gene Name:
signaling lymphocytic activation molecule family member 1
Gene Aliases:
CD150, SLAM
Chromosome Location:
Chr.38: 21653801 - 21685408 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02624257_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLAMF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003084.1 NP_001003084.1 1 - 2 96 63
GenBank mRNA AF325357.1 - 1 - 2 96 63
AF390108.1 - 1 - 2 93 63
FJ626691.1 - 1 - 2 77 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

natural killer cell differentiation
natural killer cell proliferation
leukocyte chemotaxis involved in inflammatory response
negative regulation of T cell cytokine production
positive regulation of macrophage chemotaxis
viral process
regulation of vesicle fusion
T-helper 1 cell cytokine production
positive regulation of activated T cell proliferation
innate immune response
regulation of catalytic activity
positive regulation of dendritic cell chemotaxis
signaling receptor activity
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0