This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02623312_m1
See other CD40LG GE Assays ›
Gene Symbol:
CD40LG
Gene Name:
CD40 ligand
Gene Aliases:
CD154, TNFSF5, TNLG8B
Chromosome Location:
Chr.X: 107001421 - 107023007 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02623312_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD40LG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002981.1 NP_001002981.1 1 - 2 435 109
XM_014111303.2 XP_013966778.1 3 - 4 378 109
XM_022415465.1 XP_022271173.1 2 - 3 219 109
XM_022415466.1 XP_022271174.1 3 - 4 422 109
GenBank mRNA AF086711.1 - 1 - 2 156 109
AY333790.1 - 1 - 2 435 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Inventoried Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
integrin-mediated signaling pathway
activation of JUN kinase activity
CD40 signaling pathway
platelet activation
B cell differentiation
positive regulation of interleukin-10 production
positive regulation of interleukin-12 production
positive regulation of interleukin-4 production
B cell proliferation
positive regulation of T cell proliferation
negative regulation of apoptotic process
isotype switching
immunoglobulin secretion
regulation of immunoglobulin secretion
positive regulation of NF-kappaB transcription factor activity
positive regulation of endothelial cell apoptotic process
cytokine activity
tumor necrosis factor receptor binding
CD40 receptor binding
integrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0