This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf01098017_m1
See other PHACTR4 GE Assays ›
Gene Symbol:
PHACTR4
Gene Name:
phosphatase and actin regulator 4
Gene Aliases:
-
Chromosome Location:
Chr.2: 71982524 - 72079916 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf01098017_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005617702.3 XP_005617759.1 11 - 12 2176 100
XM_005617703.3 XP_005617760.1 13 - 14 2221 100
XM_005617706.3 XP_005617763.1 12 - 13 2197 100
XM_022412502.1 XP_022268210.1 13 - 14 2186 100
XM_022412504.1 XP_022268212.1 12 - 13 2161 100
XM_022412509.1 XP_022268217.1 13 - 14 2300 100
XM_022412513.1 XP_022268221.1 13 - 14 2240 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

phosphatase modulator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0