This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf00938826_m1
See other KHDRBS3 GE Assays ›
Gene Symbol:
KHDRBS3
Gene Name:
KH RNA binding domain containing, signal transduction associated 3
Gene Aliases:
-
Chromosome Location:
Chr.13: 31397501 - 31583198 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf00938826_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KHDRBS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005627987.3 XP_005628044.1 1 - 2 135 87
XM_014118695.2 XP_013974170.1 1 - 2 135 87
XM_022426731.1 XP_022282439.1 1 - 2 140 87
XM_022426733.1 XP_022282441.1 1 - 2 137 87
XM_022426734.1 XP_022282442.1 1 - 2 139 87
XM_022426735.1 XP_022282443.1 1 - 2 138 87
XM_022426736.1 XP_022282444.1 1 - 2 135 87
XM_843255.5 XP_848348.2 1 - 2 137 87

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0