This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf00938330_m1
See other RAB3C GE Assays ›
Gene Symbol:
RAB3C
Gene Name:
RAB3C, member RAS oncogene family
Gene Aliases:
-
Chromosome Location:
Chr.2: 45459068 - 45731994 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf00938330_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAB3C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005617414.3 XP_005617471.1 2 - 3 448 113
XM_845952.4 XP_851045.3 2 - 3 505 113

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0