This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf00932673_m1
See other NDFIP1 GE Assays ›
Gene Symbol:
NDFIP1
Gene Name:
Nedd4 family interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 36978148 - 37065247 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf00932673_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDFIP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022408883.1 XP_022264591.1 575 97
XM_022408887.1 XP_022264595.1 575 97
XM_022408892.1 XP_022264600.1 557 97
XM_022408895.1 XP_022264603.1 467 97
XM_022408896.1 XP_022264604.1 467 97
XM_022408899.1 XP_022264607.1 467 97
XM_022408902.1 XP_022264610.1 455 97

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0