This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf00932626_m1
See other PPP2R2B GE Assays ›
Gene Symbol:
PPP2R2B
Gene Name:
protein phosphatase 2 regulatory subunit Bbeta
Gene Aliases:
-
Chromosome Location:
Chr.2: 41005259 - 41447048 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf00932626_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003433780.4 XP_003433828.1 4 - 5 585 109
XM_003433781.4 XP_003433829.1 2 - 3 360 109
XM_003433782.4 XP_003433830.1 3 - 4 390 109
XM_003638915.4 XP_003638963.1 3 - 4 554 109
XM_005617353.3 XP_005617410.1 3 - 4 360 109
XM_005617354.3 XP_005617411.2 2 - 3 160 109
XM_005617355.3 XP_005617412.1 4 - 5 474 109
XM_005617357.3 XP_005617414.1 3 - 4 310 109
XM_005617360.3 XP_005617417.1 3 - 4 437 109
XM_022409181.1 XP_022264889.1 3 - 4 305 109
XM_022409183.1 XP_022264891.1 4 - 5 479 109
XM_022409200.1 XP_022264908.1 3 - 4 269 109
XM_022409203.1 XP_022264911.1 3 - 4 288 109
XM_022409211.1 XP_022264919.1 3 - 4 195 109
XM_022409217.1 XP_022264925.1 2 - 3 130 109
XM_022409223.1 XP_022264931.1 2 - 3 124 109

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0