This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf00443062_g1
See other NR4A2 GE Assays ›
Gene Symbol:
NR4A2
Gene Name:
nuclear receptor subfamily 4 group A member 2
Gene Aliases:
-
Chromosome Location:
Chr.36: 2546237 - 2554559 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf00443062_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NR4A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014110865.2 XP_013966340.1 7 - 8 1756 71
XM_014110866.2 XP_013966341.1 8 - 9 1689 71
XM_022414761.1 XP_022270469.1 7 - 8 1904 71
XM_535920.6 XP_535920.2 7 - 8 1793 71

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header