Assay Details
| Gene Symbol: nlp-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071707.4 | NP_504108.1 | 1 | 467 | 128 | |
More Information
Set Membership: |
|

nlp-30| Gene Symbol: nlp-30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_071707.4 | NP_504108.1 | 1 | 467 | 128 | |
Set Membership: |
|