This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02503624_g1
See other AKT-2 GE Assays ›
Gene Symbol:
akt-2
Gene Name:
protein-coding
Gene Aliases:
CELE_F28H6.1
Chromosome Location:
Chr.X: 14147700 - 14152070 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02503624_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: akt-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029441.4 NP_001024612.1 1 - 2 102 69
NM_077956.8 NP_510357.3 1 - 2 102 69
GenBank mRNA AF072381.1 - 1 - 2 102 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

immune system process
protein phosphorylation
multicellular organism development
insulin receptor signaling pathway
determination of adult lifespan
regulation of synaptic growth at neuromuscular junction
phosphorylation
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
intracellular signal transduction
innate immune response
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
phosphatidylinositol-3,4,5-trisphosphate binding
kinase activity
transferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0