This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02502404_g1
See other CHD-3 GE Assays ›
Gene Symbol:
chd-3
Gene Name:
Chromodomain-helicase-DNA-binding protein 3 homolog
Gene Aliases:
CELE_T14G8.1
Chromosome Location:
Chr.X: 12843873 - 12852601 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02502404_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: chd-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_077739.7 NP_510140.1 5 - 6 3788 71
GenBank mRNA AF308444.1 - 5 - 6 3810 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cell fate determination
chromatin organization
chromatin remodeling
DNA duplex unwinding
negative regulation of vulval development
negative regulation of Ras protein signal transduction
nucleotide binding
DNA binding
DNA helicase activity
chromatin binding
helicase activity
ATP binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0