This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02502267_m1
See other ADT-1 GE Assays ›
Gene Symbol:
adt-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C02B4.1
Chromosome Location:
Chr.X: 12678832 - 12689279 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02502267_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: adt-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001375156.2 NP_001361951.1 5 - 6 784 96
NM_077715.5 NP_510116.2 5 - 6 784 96
GenBank mRNA AB066246.1 - 5 - 6 784 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
extracellular matrix organization
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0