This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02502151_g1
See other NPR-4 GE Assays ›
Gene Symbol:
npr-4
Gene Name:
protein-coding
Gene Aliases:
CELE_C16D6.2
Chromosome Location:
Chr.X: 12536978 - 12551684 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02502151_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: npr-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307873.3 NP_001294802.1 3 - 4 321 87
NM_001307874.4 NP_001294803.1 5 - 6 500 87
NM_001313375.3 NP_001300304.1 5 - 6 493 87
NM_077700.7 NP_510101.2 3 - 4 321 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of muscle contraction
signal transduction
G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
G-protein coupled receptor activity
neuropeptide Y receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0