This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02500310_m1
See other PLC-1 GE Assays ›
Gene Symbol:
plc-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F31B12.1
Chromosome Location:
Chr.X: 10794857 - 10838650 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02500310_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: plc-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001129669.4 NP_001123141.2 2 - 3 83 62
NM_001373489.2 NP_001360637.1 2 - 3 83 62
NM_001373491.2 NP_001360639.1 2 - 3 83 62
NM_001373492.2 NP_001360635.1 2 - 3 83 62
NM_001373493.2 NP_001360636.1 2 - 3 83 62
NM_001373494.2 NP_001359720.1 2 - 3 83 62
NM_001373495.2 NP_001360638.1 2 - 3 83 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

activation of immune response
lipid metabolic process
signal transduction
small GTPase mediated signal transduction
determination of adult lifespan
lipid catabolic process
negative regulation of GTPase activity
intracellular signal transduction
positive regulation of protein import into nucleus
phosphatidylinositol-mediated signaling
defense response to Gram-positive bacterium
regulation of defense response to bacterium, incompatible interaction
1-phosphatidyl-1D-myo-inositol 4,5-bisphosphate catabolic process
phosphatidylinositol phospholipase C activity
guanyl-nucleotide exchange factor activity
GTPase inhibitor activity
phosphoric diester hydrolase activity
hydrolase activity
Ras GTPase binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0