This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02499141_m1
See other HIM-4 GE Assays ›
Gene Symbol:
him-4
Gene Name:
protein-coding
Gene Aliases:
CELE_F15G9.4
Chromosome Location:
Chr.X: 9717565 - 9753764 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02499141_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: him-4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029411.3 NP_001024582.1 17 - 18 4982 130
NM_001373449.3 NP_001359724.1 17 4921 130
NM_001373450.2 NP_001360633.1 16 - 17 4627 130
NM_001373451.2 NP_001360634.1 16 - 17 4627 130
NM_077234.7 NP_509635.2 17 - 18 4921 130
GenBank mRNA AF074901.1 - 17 - 18 4921 130

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
basement membrane disassembly
basement membrane organization
extracellular matrix structural constituent
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0