This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02497426_g1
See other PCYT-1 GE Assays ›
Gene Symbol:
pcyt-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F08C6.2
Chromosome Location:
Chr.X: 7572110 - 7581055 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02497426_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pcyt-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001038450.4 NP_001033539.1 3 - 4 592 116
NM_001038451.3 NP_001033540.1 2 - 3 325 116

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phosphatidylcholine biosynthetic process
CDP-choline pathway
phospholipid biosynthetic process
biosynthetic process
catalytic activity
choline-phosphate cytidylyltransferase activity
transferase activity
nucleotidyltransferase activity
phosphatidylcholine binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0