This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02495632_m1
See other RGS-6 GE Assays ›
Gene Symbol:
rgs-6
Gene Name:
protein-coding
Gene Aliases:
CELE_C41G11.3
Chromosome Location:
Chr.X: 5720381 - 5733778 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02495632_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: rgs-6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307891.3 NP_001294820.1 3 - 4 359 65
NM_001307892.4 NP_001294821.1 1 - 2 516 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

negative regulation of signal transduction

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header