This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02495424_g1
See other DPYD-1 GE Assays ›
Gene Symbol:
dpyd-1
Gene Name:
Dihydropyrimidine dehydrogenase [NADP(+)]
Gene Aliases:
CELE_C25F6.3
Chromosome Location:
Chr.X: 5467870 - 5472684 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02495424_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: dpyd-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_076526.6 NP_508927.2 11 - 12 2879 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

thymine catabolic process
uracil catabolic process
thymidine catabolic process
beta-alanine biosynthetic process
oxidation-reduction process
nucleotide binding
uracil binding
catalytic activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
dihydropyrimidine dehydrogenase (NADP+) activity
metal ion binding
NADP binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0