This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02494442_g1
See other PDI-2 GE Assays ›
Gene Symbol:
pdi-2
Gene Name:
Protein disulfide-isomerase 2
Gene Aliases:
CELE_C07A12.4
Chromosome Location:
Chr.X: 4523733 - 4532847 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02494442_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pdi-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_076377.5 NP_508778.1 3 - 4 958 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein folding
peptidyl-proline hydroxylation to 4-hydroxy-L-proline
endoplasmic reticulum unfolded protein response
response to endoplasmic reticulum stress
IRE1-mediated unfolded protein response
macromolecule modification
cell redox homeostasis
oxidation-reduction process
protein deglutathionylation
extracellular matrix assembly
protein disulfide isomerase activity
protein-glutamine gamma-glutamyltransferase activity
isomerase activity
peptidyl-proline 4-dioxygenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0