This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02484751_m1
See other INTS-2 GE Assays ›
Gene Symbol:
ints-2
Gene Name:
protein-coding
Gene Aliases:
CELE_ZC376.6
Chromosome Location:
Chr.V: 14185686 - 14192161 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02484751_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ints-2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001269640.4 NP_001256569.1 14 - 15 2980 114
NM_001269641.2 NP_001256570.1 14 - 15 2973 114

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s)

snRNA 3'-end processing

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0