This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02479267_g1
See other C25E10.8 GE Assays ›
Gene Symbol:
C25E10.8 swm-1
Gene Name:
Uncharacterized protein
Serine protease inhibitor swm-1
Gene Aliases:
CELE_C25E10.8
CELE_C25E10.9
Chromosome Location:
Chr.V: 9053262 - 9054783 on Build WBcel235
Chromosome Location:
Chr.V: 9053318 - 9054910 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02479267_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C25E10.8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383365.1 6 - 7 791 84
Gene Symbol: swm-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_072945.3 NP_505346.1 4 - 5 1064 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

spermatogenesis
spermatid development
negative regulation of peptidase activity
negative regulation of endopeptidase activity
cell differentiation
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0