This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02478585_g1
See other PXN-1 GE Assays ›
Gene Symbol:
pxn-1
Gene Name:
Peroxidasin homolog pxn-1
Gene Aliases:
CELE_ZK994.3
Chromosome Location:
Chr.V: 8494889 - 8502442 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02478585_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: pxn-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_072787.4 NP_505188.3 15 - 16 3146 130

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
embryonic body morphogenesis
muscle attachment
hydrogen peroxide catabolic process
regulation of axon regeneration
oxidation-reduction process
cellular oxidant detoxification
positive regulation of axon guidance
peroxidase activity
oxidoreductase activity
heme binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0