This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02470079_m1
See other NPR-5 GE Assays ›
Gene Symbol:
npr-5 nlp-70
Gene Name:
protein-coding
Neuropeptide-Like Protein
Gene Aliases:
CELE_Y58G8A.4
CELE_Y58G8A.5
Chromosome Location:
Chr.V: 276984 - 281716 on Build WBcel235
Chromosome Location:
Chr.V: 271921 - 281718 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02470079_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: npr-5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001029093.5 NP_001024264.1 1 - 2 156 76
NM_001029094.4 NP_001024265.1 1 - 2 156 76
Gene Symbol: nlp-70
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001380588.1 NP_001367810.1 1 - 2 158 76

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001380588.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of muscle contraction
G-protein coupled receptor signaling pathway
neuropeptide signaling pathway
G-protein coupled receptor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header