This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ce02469396_m1
See other EGL-8 GE Assays ›
Gene Symbol:
egl-8
Gene Name:
protein-coding
Gene Aliases:
CELE_B0348.4
Chromosome Location:
Chr.V: 20818 - 55451 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID
Ce02469396_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: egl-8
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001026175.4
NP_001021346.1
16 - 17
3951
68
NM_001026176.3
NP_001021347.1
17 - 18
3987
68
NM_001313085.3
NP_001300014.1
18
4048
68
NM_001313087.3
NP_001300016.1
17 - 18
3912
68
NM_001313089.4
NP_001300018.1
17
3676
68
NM_001313091.3
NP_001300020.1
17
3738
68
NM_001313093.3
NP_001300022.1
16
3495
68
NM_001313095.4
NP_001300024.1
17
4099
68
NM_001313097.3
NP_001300026.1
16
3721
68
NM_001313099.3
NP_001300028.1
16
3646
68
NM_001313101.3
NP_001300030.1
15
3403
68
NM_001313103.4
NP_001300032.1
15 - 16
3648
68
NM_001313105.3
NP_001300034.1
14
3223
68
NM_001372980.2
NP_001360389.1
16
3639
68
NM_001372988.2
NP_001360397.1
14
3375
68
NM_001380576.2
NP_001368552.1
17 - 18
4012
68
NM_001380577.1
NP_001368553.1
16 - 17
3882
68
NM_001380578.2
NP_001368554.1
15 - 16
3700
68
NM_001380579.1
NP_001368555.1
14 - 15
3459
68
NM_001380580.2
NP_001368556.1
16 - 17
4112
68
NM_001380581.2
NP_001368557.1
15 - 16
3873
68
NM_001380582.2
NP_001368558.1
15 - 16
3780
68
NM_001380583.1
NP_001368559.1
14 - 15
3530
68
NM_001380584.1
NP_001368560.1
14 - 15
3430
68
GenBank mRNA
AF179426.1
-
17 - 18
3987
68
Back To Top
More Information
Set Membership:
Amplicon length less than or equal to 70
Caenorhabditis elegans
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
chemotaxis
signal transduction
dopamine receptor signaling pathway
positive regulation of acetylcholine secretion, neurotransmission
lipid catabolic process
protein localization to synapse
intracellular signal transduction
regulation of locomotion
positive regulation of locomotion
regulation of pharyngeal pumping
thermotaxis
regulation of oviposition
phosphatidylinositol-mediated signaling
positive regulation of axon regeneration
phosphatidylinositol phospholipase C activity
calcium ion binding
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0