This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02466805_s1
See other JC8.4 GE Assays ›
Gene Symbol:
JC8.4 cox-11
Gene Name:
CMP/dCMP-type deaminase domain-containing protein
protein-coding
Gene Aliases:
CELE_JC8.4
CELE_JC8.5
Chromosome Location:
Chr.IV: 13243675 - 13244205 on Build WBcel235
Chromosome Location:
Chr.IV: 13243673 - 13247161 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
-
Assay ID Ce02466805_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: JC8.4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_070145.3 NP_502546.1 1 440 86
Gene Symbol: cox-11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_070146.6 NP_502547.2 1 442 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_070146.6 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

tRNA wobble adenosine to inosine editing
catalytic activity
tRNA-specific adenosine-34 deaminase activity
copper ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0