Assay Details
| Gene Symbol: dpy-26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_069977.8 | NP_502378.1 | 11 | 4785 | 70 | |
| Gene Symbol: C25G4.3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268715.3 | NP_001255644.1 | 2 | 631 | 70 | |
| NM_001268716.3 | NP_001255645.1 | 1 | 469 | 70 | ||
More Information
Set Membership: |
|

dpy-26
Genomic Map