This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02464700_g1
See other FAN-1 GE Assays ›
Gene Symbol:
fan-1
Gene Name:
protein-coding
Gene Aliases:
CELE_C01G5.8
Chromosome Location:
Chr.IV: 6548685 - 6552511 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02464700_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fan-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001383106.1 13 - 14 2043 100
NM_068596.5 NP_500997.1 12 - 13 2260 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
cellular response to DNA damage stimulus
interstrand cross-link repair
nucleic acid phosphodiester bond hydrolysis
nucleic acid binding
nuclease activity
phosphodiesterase I activity
protein binding
5'-3' exonuclease activity
hydrolase activity
hydrolase activity, acting on ester bonds
5'-flap endonuclease activity
metal ion binding
flap-structured DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0