This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02464348_m1
See other UNC-77 GE Assays ›
Gene Symbol:
unc-77
Gene Name:
protein-coding
Gene Aliases:
CELE_C11D2.6
Chromosome Location:
Chr.IV: 6157306 - 6183857 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02464348_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001306892.3 NP_001293821.1 13 - 14 2269 96
NM_001306893.3 NP_001293822.1 13 - 14 2269 96
NM_001306896.3 NP_001293825.1 14 - 15 2323 96
NM_001306897.3 NP_001293826.1 14 - 15 2323 96
NM_001307702.3 NP_001294631.1 13 - 14 2305 96
NM_001307703.3 NP_001294632.1 12 - 13 2125 96
NM_068525.4 NP_500926.3 13 - 14 2408 96
NM_068526.5 NP_500927.3 12 - 13 2125 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

ion transport
positive regulation of synaptic transmission, cholinergic
positive regulation of synaptic transmission, GABAergic
ion transmembrane transport
positive regulation of locomotion
transmembrane transport
cation transmembrane transport
ion channel activity
cation channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0