This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02460565_m1
See other PLX-1 GE Assays ›
Gene Symbol:
plx-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y55F3AL.1
Chromosome Location:
Chr.IV: 959521 - 984395 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02460565_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: plx-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307625.3 NP_001294554.1 19 - 20 4429 81
NM_001307626.3 NP_001294555.1 16 - 17 3862 81
NM_001307627.3 NP_001294556.1 16 - 17 3862 81
NM_001307628.3 NP_001294557.1 7 - 8 2116 81
NM_001307629.3 NP_001294558.1 7 - 8 2116 81
NM_067617.6 NP_500018.3 19 - 20 4429 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cell adhesion
signal transduction
small GTPase mediated signal transduction
regulation of cell shape
epidermis development
regulation of cell migration
regulation of GTPase activity
nematode male tail tip morphogenesis
positive regulation of axonogenesis
regulation of cellular component movement
semaphorin-plexin signaling pathway
nematode male tail mating organ morphogenesis
semaphorin-plexin signaling pathway involved in axon guidance
protein binding
semaphorin receptor activity
Rac GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0