This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02458251_g1
See other FAT-3 GE Assays ›
Gene Symbol:
fat-3
Gene Name:
protein-coding
Gene Aliases:
CELE_W08D2.4
Chromosome Location:
Chr.IV: 9803023 - 9806119 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02458251_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: fat-3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001268497.2 NP_001255426.1 4 - 5 780 74
NM_001307156.3 NP_001294085.1 2 - 3 437 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

serotonin secretion
lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
unsaturated fatty acid biosynthetic process
muscle contraction
protein localization
acetylcholine secretion, neurotransmission
oviposition
defecation
endocytic recycling
multicellular organism growth
collagen and cuticulin-based cuticle development
locomotion
pharyngeal pumping
regulation of innate immune response
oxidation-reduction process
stearoyl-CoA 9-desaturase activity
oxidoreductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0