This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02455891_g1
See other ITR-1 GE Assays ›
Gene Symbol:
itr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F33D4.2
Chromosome Location:
Chr.IV: 7675174 - 7696930 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02455891_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: itr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027999.4 NP_001023170.1 8 - 9 1286 75
NM_001028000.5 NP_001023171.1 6 - 7 1118 75
NM_001028002.4 NP_001023173.1 7 - 8 1214 75
NM_001268376.4 NP_001255305.1 7 - 8 1148 75
NM_001380339.1 NP_001367466.1 7 - 8 1148 75
GenBank mRNA AF168688.1 - 7 - 8 1214 75
AJ243179.2 - 7 - 8 1236 75
AJ243180.2 - 8 - 9 1309 75
AJ243181.2 - 7 - 8 1229 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

oocyte maturation
ion transport
calcium ion transport
muscle contraction
positive regulation of cytosolic calcium ion concentration
multicellular organism development
rhythmic behavior
programmed cell death
regulation of cell migration
defecation
sperm ejaculation
regulation of pharyngeal pumping
inositol phosphate-mediated signaling
inositol lipid-mediated signaling
embryonic morphogenesis
multicellular organismal reproductive process
hermaphrodite genitalia morphogenesis
release of sequestered calcium ion into cytosol
regulation of filopodium assembly
transmembrane transport
male mating behavior
calcium ion transmembrane transport
positive regulation of pharyngeal pumping
ion channel activity
inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
calcium channel activity
calcium ion binding
protein binding
myosin binding
phosphatidylinositol binding
inositol 1,4,5 trisphosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0