This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02455870_m1
See other ITR-1 GE Assays ›
Gene Symbol:
itr-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F33D4.2
Chromosome Location:
Chr.IV: 7675174 - 7696930 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02455870_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: itr-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027999.4 NP_001023170.1 19 - 20 5188 69
NM_001028000.5 NP_001023171.1 17 - 18 5020 69
NM_001028001.5 NP_001023172.1 19 - 20 5145 69
NM_001028002.4 NP_001023173.1 18 - 19 5116 69
GenBank mRNA AF168688.1 - 18 - 19 5116 69
AJ243179.2 - 18 - 19 5138 69
AJ243180.2 - 19 - 20 5211 69
AJ243181.2 - 18 - 19 5131 69
AJ243182.2 - 19 - 20 5165 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

oocyte maturation
ion transport
calcium ion transport
muscle contraction
positive regulation of cytosolic calcium ion concentration
multicellular organism development
rhythmic behavior
programmed cell death
regulation of cell migration
defecation
sperm ejaculation
regulation of pharyngeal pumping
inositol phosphate-mediated signaling
inositol lipid-mediated signaling
embryonic morphogenesis
multicellular organismal reproductive process
hermaphrodite genitalia morphogenesis
release of sequestered calcium ion into cytosol
regulation of filopodium assembly
transmembrane transport
male mating behavior
calcium ion transmembrane transport
positive regulation of pharyngeal pumping
ion channel activity
inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
calcium channel activity
calcium ion binding
protein binding
myosin binding
phosphatidylinositol binding
inositol 1,4,5 trisphosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0