This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02455752_m1
See other CLA-1 GE Assays ›
Gene Symbol:
cla-1
Gene Name:
protein-coding
Gene Aliases:
CELE_F45E4.3
Chromosome Location:
Chr.IV: 7588931 - 7628238 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02455752_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: cla-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307215.3 NP_001294144.1 18 - 19 6700 86
NM_001307219.4 NP_001294148.2 31 - 32 25540 86
NM_001383125.1 11 - 12 6030 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
synapse assembly
regulation of dopamine secretion
synaptic vesicle exocytosis
vesicle-mediated transport
calcium ion regulated exocytosis
regulation of calcium ion-dependent exocytosis
synaptic vesicle endocytosis
maintenance of presynaptic active zone structure
calcium ion-regulated exocytosis of neurotransmitter
cellular response to calcium ion
synaptic vesicle clustering
spontaneous synaptic transmission
SNARE binding
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
Rab GTPase binding
syntaxin binding
clathrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0