This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02455506_m1
See other UNC-82 GE Assays ›
Gene Symbol:
unc-82
Gene Name:
protein-coding
Gene Aliases:
CELE_B0496.3
Chromosome Location:
Chr.IV: 7424498 - 7444664 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02455506_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: unc-82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001268365.3 NP_001255294.1 12 - 13 2172 62
NM_001268366.3 NP_001255295.1 7 - 8 1453 62
NM_001268369.2 NP_001255298.1 9 - 10 1558 62
NM_001307278.3 NP_001294207.1 7 - 8 1504 62
NM_001372869.1 NP_001359805.1 9 - 10 1558 62
NM_001380324.1 NP_001367366.1 9 - 10 1558 62
NM_001380325.1 NP_001367367.1 12 - 13 1922 62
NM_068785.7 NP_501186.5 7 - 8 1453 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
phosphorylation
intracellular signal transduction
cellular response to glucose starvation
striated muscle paramyosin thick filament assembly
striated muscle myosin thick filament assembly
protein kinase activity
protein serine/threonine kinase activity
ATP binding
structural constituent of muscle
kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0