This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02453428_g1
See other Y79H2A.3 GE Assays ›
Gene Symbol:
Y79H2A.3
Gene Name:
protein-coding
Gene Aliases:
CELE_Y79H2A.3
Chromosome Location:
Chr.III: 12031516 - 12052306 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02453428_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Y79H2A.3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027784.5 NP_001022955.2 1 - 2 126 87
NM_001027785.5 NP_001022956.2 1 - 2 116 87
NM_001268226.3 NP_001255155.1 1 - 2 116 87
NM_001268227.4 NP_001255156.1 1 - 2 129 87
NM_001268229.3 NP_001255158.1 1 - 2 116 87
NM_001268230.2 NP_001255159.1 1 - 2 126 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0