This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ce02453176_m1
See other CRH-1 GE Assays ›
Gene Symbol:
crh-1
Gene Name:
protein-coding
Gene Aliases:
CELE_Y41C4A.4
Chromosome Location:
Chr.III: 11657738 - 11695165 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID Ce02453176_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: crh-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001027688.5 NP_001022859.1 4 - 5 551 102
NM_001027689.3 NP_001022860.1 4 - 5 455 102
NM_001027690.3 NP_001022861.1 4 - 5 445 102
NM_001027691.4 NP_001022862.1 4 - 5 567 102
NM_001027692.4 NP_001022863.1 4 - 5 452 102
GenBank mRNA AB081595.1 - 4 - 5 452 102
AB081596.1 - 4 - 5 534 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
determination of adult lifespan
intracellular signal transduction
thermosensory behavior
positive regulation of transcription from RNA polymerase II promoter
transcription coactivator binding
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header